Product Details

SNP ID
rs75574634
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:176471451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATTATAAAATAAAACACAAAT[A/G]TAGAAAACCAAACAAAACAAGTGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAPPA2 PubMed Links

Gene Details

Gene
PAPPA2
Gene Name
pappalysin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020318.2 Intron NP_064714.2
NM_021936.2 Intron NP_068755.2
XM_005245422.3 Intron XP_005245479.1
XM_011509857.1 Intron XP_011508159.1
XM_011509858.2 Intron XP_011508160.1
XM_017002023.1 Intron XP_016857512.1
XM_017002024.1 Intron XP_016857513.1
XM_017002025.1 Intron XP_016857514.1

View Full Product Details