Product Details

SNP ID
rs77734316
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:179960942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAAAAAAATAACTGGCATACTG[A/G]GGGGTCTGTGAACTGAGAAAGTTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP350 PubMed Links

Gene Details

Gene
CEP350
Gene Name
centrosomal protein 350
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014810.4 Intron NP_055625.4
XM_011510191.2 Intron XP_011508493.1
XM_011510198.2 Intron XP_011508500.1
XM_017002929.1 Intron XP_016858418.1
XM_017002930.1 Intron XP_016858419.1
XM_017002931.1 Intron XP_016858420.1
Gene
LOC101928933
Gene Name
uncharacterized LOC101928933
There are no transcripts associated with this gene.

View Full Product Details