Product Details

SNP ID
rs80053700
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:184404824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAAACAGTCACTCTCTACCCC[A/C]TCGTGATGCGTCCTTTCATAGATTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf21 PubMed Links

Gene Details

Gene
C1orf21
Gene Name
chromosome 1 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030806.3 Intron NP_110433.1

View Full Product Details