Product Details

SNP ID
rs78194024
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:204032679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTTGCTATTAAAAAAAAAAAT[C/G]TCAACTTATTTCATCTCCTACTAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details