Product Details

SNP ID
rs78444759
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:155865983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTATAGGCCTAATCTGGACCTTTA[A/C]AGCCTAAACCCAATTTAGCCTCAAT
Phenotype
MIM: 610393 MIM: 608741
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GON4L PubMed Links

Gene Details

Gene
GON4L
Gene Name
gon-4 like
There are no transcripts associated with this gene.

Gene
SYT11
Gene Name
synaptotagmin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152280.4 Intron NP_689493.3
XM_005245014.2 Intron XP_005245071.1
XM_017000759.1 Intron XP_016856248.1

View Full Product Details