Product Details

SNP ID
rs75854315
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:156253265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTAAAGAGGGAGAGACAGAAAT[A/G]AAGGCAGAAAGGAGGCAAATGGCTG
Phenotype
MIM: 112260 MIM: 614579 MIM: 610962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323614.1 Intron NP_001310543.1
NM_001323615.1 Intron NP_001310544.1
NM_001323616.1 Intron NP_001310545.1
NM_001323617.1 Intron NP_001310546.1
NM_015327.2 Intron NP_056142.2
XM_017000842.1 Intron XP_016856331.1
XM_017000843.1 Intron XP_016856332.1

View Full Product Details