Product Details

SNP ID
rs77401325
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:157793058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCTGCTTTTGGGCTTTTTGGA[A/C]AAATAAACTCACAGACCCTATCAAA
Phenotype
MIM: 606508
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FCRL1 PubMed Links

Gene Details

Gene
FCRL1
Gene Name
Fc receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159397.1 4161 Intron NP_001152869.1
NM_001159398.1 4161 Intron NP_001152870.1
NM_052938.4 4161 Intron NP_443170.1
XM_005244866.2 4161 UTR 3 XP_005244923.1
XM_005244867.2 4161 UTR 3 XP_005244924.1
XM_005244869.3 4161 UTR 3 XP_005244926.1
XM_011509125.1 4161 UTR 3 XP_011507427.1
XM_011509126.1 4161 UTR 3 XP_011507428.1
XM_011509127.2 4161 UTR 3 XP_011507429.1
XM_011509128.1 4161 UTR 3 XP_011507430.1
XM_011509129.1 4161 UTR 3 XP_011507431.1
XM_011509130.1 4161 UTR 3 XP_011507432.1
XM_011509131.1 4161 Intron XP_011507433.1
XM_011509132.2 4161 Intron XP_011507434.1
XM_011509133.2 4161 UTR 3 XP_011507435.1
XM_011509134.2 4161 UTR 3 XP_011507436.1
XM_011509135.2 4161 UTR 3 XP_011507437.1
XM_011509136.1 4161 Intron XP_011507438.1
XM_011509137.2 4161 UTR 3 XP_011507439.1
XM_017000227.1 4161 UTR 3 XP_016855716.1

View Full Product Details