Product Details

SNP ID
rs76536291
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:26022520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTCGATGGGCCTCAGTCTTCC[C/T]ATCTGTACAATGACAGCACAGGACT
Phenotype
MIM: 601738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXTL1 PubMed Links

Gene Details

Gene
EXTL1
Gene Name
exostosin like glycosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004455.2 772 UTR 5 NP_004446.2
XM_005245779.3 772 UTR 5 XP_005245836.1
XM_017000650.1 772 UTR 5 XP_016856139.1

View Full Product Details