Product Details
- SNP ID
-
rs77417309
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:26320192 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGATACAAACTGGACTCTCAGGA[A/C]AAAACGACACCAGCCAAACCAGCAG
- Phenotype
-
MIM: 114280
MIM: 609151
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AIM1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1071849,rs17645] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIM1L
- Gene Name
- absent in melanoma 1-like
There are no transcripts associated with this gene.
- Gene
- CD52
- Gene Name
- CD52 molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001803.2 |
174 |
Missense Mutation |
AAA,CAA |
K26Q |
NP_001794.2 |
- Gene
- UBXN11
- Gene Name
- UBX domain protein 11
There are no transcripts associated with this gene.
View Full Product Details