Product Details

SNP ID
rs74645064
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:27355301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCTTCCTCCAGTCGCCCCAG[A/G]TCCTGGGGCTGGTGTGTCAGAAGCT
Phenotype
MIM: 604468 MIM: 608042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K6 PubMed Links

Gene Details

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297609.1 4202 UTR 3 NP_001284538.1
NM_004672.4 4202 UTR 3 NP_004663.3
XM_017002771.1 4202 UTR 3 XP_016858260.1
XM_017002772.1 4202 Intron XP_016858261.1
Gene
SYTL1
Gene Name
synaptotagmin like 1
There are no transcripts associated with this gene.

View Full Product Details