Product Details

SNP ID
rs78247007
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:15440505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTCCTGCAGATCTCCCTCCAG[C/T]ACCTCAAGAACGACACGTGGAGGCA
Phenotype
MIM: 601405
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTRC PubMed Links

Gene Details

Gene
CTRC
Gene Name
chymotrypsin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007272.2 168 Missense Mutation CAC,TAC H49Y NP_009203.2
XM_011540550.1 168 Missense Mutation CAC,TAC H49Y XP_011538852.1

View Full Product Details