Product Details
- SNP ID
-
rs76164149
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:18481093 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGTGACTGTGGCCTACAGGCTG[G/T]ACAAGTCGAGGCCTGCCCCAGCCCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLHDC7A
PubMed Links
Gene Details
- Gene
- KLHDC7A
- Gene Name
- kelch domain containing 7A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152375.2 |
164 |
Missense Mutation |
GAC,TAC |
D38Y |
NP_689588.2 |
View Full Product Details