Product Details

SNP ID
rs76808404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233477381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTCATCGGGAGTAGAGCCGTG[C/T]AGCGGCGCGGTTATCTGAAGCTCCC
Phenotype
MIM: 601826 MIM: 610570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKD PubMed Links

Gene Details

Gene
DGKD
Gene Name
diacylglycerol kinase delta
There are no transcripts associated with this gene.

Gene
USP40
Gene Name
ubiquitin specific peptidase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018218.2 3847 UTR 3 NP_060688.1
XM_006712612.3 3847 UTR 3 XP_006712675.1
XM_011511396.2 3847 UTR 3 XP_011509698.1
XM_011511397.2 3847 UTR 3 XP_011509699.1
XM_011511398.2 3847 Intron XP_011509700.1
XM_011511399.2 3847 UTR 3 XP_011509701.1
XM_011511400.2 3847 UTR 3 XP_011509702.1
XM_011511401.2 3847 Intron XP_011509703.1
XM_017004426.1 3847 UTR 3 XP_016859915.1
XM_017004427.1 3847 UTR 3 XP_016859916.1
XM_017004428.1 3847 UTR 3 XP_016859917.1

View Full Product Details