Product Details

SNP ID
rs78408678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8731968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGAGACTCGAAAGACTTGGGGT[A/T]CTGCGAGTTTGTGACTGTGAAGACA
Phenotype
MIM: 615759
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIDINS220 PubMed Links

Gene Details

Gene
KIDINS220
Gene Name
kinase D-interacting substrate 220kDa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020738.2 4250 Missense Mutation AGA,AGT R1356S NP_065789.1

View Full Product Details