Product Details

SNP ID
rs76059450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10046351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTCTCTGACTCTGGGGATGTC[A/C]CCACCACTGTGCATATGGATGCAGC
Phenotype
MIM: 603301
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLF11 PubMed Links

Gene Details

Gene
KLF11
Gene Name
Kruppel like factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177716.1 261 Missense Mutation ACC,CCC T65P NP_001171187.1
NM_001177718.1 261 Missense Mutation ACC,CCC T65P NP_001171189.1
NM_003597.4 261 Missense Mutation ACC,CCC T82P NP_003588.1
Gene
LOC101929882
Gene Name
uncharacterized LOC101929882
There are no transcripts associated with this gene.

View Full Product Details