Product Details

SNP ID
rs77836288
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:167956631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCCTAATTAATAAGTTATTTC[A/G]TTTTTCTTGTGACCTCTCCCCCTTG
Phenotype
MIM: 607648
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
STK39 PubMed Links

Gene Details

Gene
STK39
Gene Name
serine/threonine kinase 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013233.2 Intron NP_037365.2
XM_005246465.2 Intron XP_005246522.1
XM_017003813.1 Intron XP_016859302.1
XM_017003814.1 Intron XP_016859303.1
XM_017003815.1 Intron XP_016859304.1
XM_017003816.1 Intron XP_016859305.1
XM_017003817.1 Intron XP_016859306.1

View Full Product Details