Product Details

SNP ID
rs76148228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197453925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAAGCGACTTTGGAAGCAATTTG[G/T]CCATTGGTGCCTTTGGGCTCTTCCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COQ10B PubMed Links

Gene Details

Gene
COQ10B
Gene Name
coenzyme Q10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320818.1 162 Intron NP_001307747.1
NM_001320819.1 162 UTR 5 NP_001307748.1
NM_001320820.1 162 UTR 5 NP_001307749.1
NM_025147.4 162 Intron NP_079423.1

View Full Product Details