Product Details

SNP ID
rs80282360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202275121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACATTTTATTACAGGTTCTAAA[C/T]GAGAAGAAACTTCAAGAGGTTGATA
Phenotype
MIM: 616742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOP58 PubMed Links

Gene Details

Gene
NOP58
Gene Name
NOP58 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015934.4 280 Silent Mutation AAC,AAT N18N NP_057018.1
Gene
SNORD70
Gene Name
small nucleolar RNA, C/D box 70
There are no transcripts associated with this gene.

View Full Product Details