Product Details

SNP ID
rs76854461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207823872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGCACCCATTAAAACGCACTA[A/C]GGAAAGACCCTTGAGTCAGAGATTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHM3 PubMed Links

Gene Details

Gene
PLEKHM3
Gene Name
pleckstrin homology domain containing M3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080475.2 7223 UTR 3 NP_001073944.1
XM_005246545.3 7223 UTR 3 XP_005246602.1
XM_006712521.3 7223 Intron XP_006712584.1
XM_011511160.2 7223 UTR 3 XP_011509462.1
XM_011511163.2 7223 Intron XP_011509465.1
XM_017004071.1 7223 Intron XP_016859560.1
XM_017004072.1 7223 UTR 3 XP_016859561.1
XM_017004073.1 7223 UTR 3 XP_016859562.1

View Full Product Details