Product Details

SNP ID
rs75520497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:210188033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTTATTATTTACATTGCACTTA[C/T]GCTTGAAATTATCTCTTTTACATAT
Phenotype
MIM: 609576
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACADL PubMed Links

Gene Details

Gene
ACADL
Gene Name
acyl-CoA dehydrogenase, long chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001608.3 2877 UTR 3 NP_001599.1
XM_005246517.4 2877 UTR 3 XP_005246574.1
XM_017003955.1 2877 UTR 3 XP_016859444.1

View Full Product Details