Product Details

SNP ID
rs79070675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:229025143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCATACAGAGGAGGGCATCTGC[C/T]GTGGGAGAGGAACAGAGGGCCCTCC
Phenotype
MIM: 612930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PID1 PubMed Links

Gene Details

Gene
PID1
Gene Name
phosphotyrosine interaction domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100818.1 1441 UTR 3 NP_001094288.1
NM_017933.4 1441 UTR 3 NP_060403.3
XM_011511384.2 1441 UTR 3 XP_011509686.1
XM_017004404.1 1441 UTR 3 XP_016859893.1
XM_017004405.1 1441 UTR 3 XP_016859894.1
XM_017004406.1 1441 UTR 3 XP_016859895.1
XM_017004407.1 1441 UTR 3 XP_016859896.1

View Full Product Details