Product Details

SNP ID
rs37822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3435034 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCCAGTATTTATCTATAATAGCA[A/G]TATCTAAAGATCTATATATTATCAA
Phenotype
MIM: 614246 MIM: 614685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAA60 PubMed Links

Gene Details

Gene
NAA60
Gene Name
N(alpha)-acetyltransferase 60, NatF catalytic subunit
There are no transcripts associated with this gene.

Gene
ZNF597
Gene Name
zinc finger protein 597
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152457.2 2896 UTR 3 NP_689670.1

View Full Product Details