Product Details

SNP ID
rs39728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3403076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGATAGGGTCACCCTGTTGGGT[A/T]GCAGTTGGACTTTGGGGAACTTCAT
Phenotype
MIM: 603900
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF174 PubMed Links

Gene Details

Gene
ZNF174
Gene Name
zinc finger protein 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032292.2 Intron NP_001027463.1
NM_003450.2 Intron NP_003441.1
Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
There are no transcripts associated with this gene.

View Full Product Details