Product Details

SNP ID
rs237796
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:229321461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCCATATCCACCCTATAGTACA[A/C]AAATTCATTTCAAATGCAAACGTTG
Phenotype
MIM: 616762
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCSAP PubMed Links

Gene Details

Gene
CCSAP
Gene Name
centriole, cilia and spindle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145257.4 4665 UTR 3 NP_660300.3

View Full Product Details