Product Details
- SNP ID
-
rs161557
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:143820488 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCATTCTTTTCTATAGGTTCTCTG[C/T]ATGTTTGGAAGTCGGAATTGGTTGA
- Phenotype
-
MIM: 609961
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HMHB1
PubMed Links
Gene Details
- Gene
- HMHB1
- Gene Name
- histocompatibility minor HB-1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021182.1 |
153 |
Missense Mutation |
CAT,TAT |
H16Y |
NP_067005.1 |
View Full Product Details