Product Details

SNP ID
rs2032668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:13325453 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGAATGAAAAAGTTGGGTGACAC[A/C]AACTCTTCAGAAGGAAAAATACATA
Phenotype
MIM: 400009
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UTY PubMed Links

Gene Details

Gene
UTY
Gene Name
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258249.1 Intron NP_001245178.1
NM_001258250.1 Intron NP_001245179.1
NM_001258251.1 Intron NP_001245180.1
NM_001258252.1 Intron NP_001245181.1
NM_001258253.1 Intron NP_001245182.1
NM_001258254.1 Intron NP_001245183.1
NM_001258255.1 Intron NP_001245184.1
NM_001258256.1 Intron NP_001245185.1
NM_001258257.1 Intron NP_001245186.1
NM_001258258.1 Intron NP_001245187.1
NM_001258259.1 Intron NP_001245188.1
NM_001258260.1 Intron NP_001245189.1
NM_001258261.1 Intron NP_001245190.1
NM_001258262.1 Intron NP_001245191.1
NM_001258263.1 Intron NP_001245192.1
NM_001258264.1 Intron NP_001245193.1
NM_001258265.1 Intron NP_001245194.1
NM_001258266.1 Intron NP_001245195.1
NM_001258267.1 Intron NP_001245196.1
NM_001258268.1 Intron NP_001245197.1
NM_001258269.1 Intron NP_001245198.1
NM_001258270.1 Intron NP_001245199.1
NM_007125.4 Intron NP_009056.3
NM_182659.1 Intron NP_872600.1
NM_182660.1 Intron NP_872601.1
XM_005262518.3 Intron XP_005262575.1
XM_006724875.3 Intron XP_006724938.1
XM_011531441.2 Intron XP_011529743.1
XM_011531442.2 Intron XP_011529744.1
XM_011531443.2 Intron XP_011529745.1
XM_011531445.2 Intron XP_011529747.1
XM_011531446.2 Intron XP_011529748.1
XM_011531447.2 Intron XP_011529749.1
XM_011531448.2 Intron XP_011529750.1
XM_011531449.2 Intron XP_011529751.1
XM_011531450.2 Intron XP_011529752.1
XM_011531451.2 Intron XP_011529753.1
XM_011531452.2 Intron XP_011529754.1
XM_011531453.2 Intron XP_011529755.1
XM_011531454.2 Intron XP_011529756.1
XM_011531455.2 Intron XP_011529757.1
XM_011531457.2 Intron XP_011529759.1
XM_011531458.2 Intron XP_011529760.1
XM_011531459.2 Intron XP_011529761.1
XM_011531460.2 Intron XP_011529762.1
XM_011531461.2 Intron XP_011529763.1
XM_011531462.2 Intron XP_011529764.1
XM_011531463.2 Intron XP_011529765.1
XM_011531464.2 Intron XP_011529766.1
XM_011531465.2 Intron XP_011529767.1
XM_017030066.1 Intron XP_016885555.1
XM_017030067.1 Intron XP_016885556.1
XM_017030068.1 Intron XP_016885557.1
XM_017030069.1 Intron XP_016885558.1
XM_017030070.1 Intron XP_016885559.1
XM_017030071.1 Intron XP_016885560.1
XM_017030072.1 Intron XP_016885561.1
XM_017030073.1 Intron XP_016885562.1
XM_017030074.1 Intron XP_016885563.1

View Full Product Details