Product Details

SNP ID
rs2032621
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19710852 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGCCCCTGGATGGTTTAAGTTA[T/C]ATTTTCATCAGTCATCCAGTTAAGA
Phenotype
MIM: 426000
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 Intron NP_001140177.1
NM_001146706.1 Intron NP_001140178.1
NM_004653.4 Intron NP_004644.2
XM_005262560.2 Intron XP_005262617.1
XM_005262561.2 Intron XP_005262618.1
XM_011531468.2 Intron XP_011529770.1

View Full Product Details