Product Details

SNP ID
rs407660
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:157184809 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAATTTTGCCCAAGCACCACCTTG[A/G]TCTTCTGAAAGCCCCTCTGGAGAAC
Phenotype
MIM: 186973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITK PubMed Links

Gene Details

Gene
ITK
Gene Name
IL2 inducible T-cell kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005546.3 Intron NP_005537.3
XM_017009443.1 Intron XP_016864932.1

View Full Product Details