Product Details

SNP ID
rs45074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127657557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCCCATCCCCTCATCCCTAGT[C/G]CCCCTTGGGAACGAATCAGCAGATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTXN3 PubMed Links

Gene Details

Gene
CTXN3
Gene Name
cortexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048252.2 610 Silent Mutation GTC,GTG V12V NP_001041717.1
NM_001127385.1 610 Silent Mutation GTC,GTG V12V NP_001120857.1

View Full Product Details