Product Details

SNP ID
rs258326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89687066 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGGCCCGGGACTTGGGAAGAGCA[A/G]GCTCCGGGATCCAGCTCGGGCGCTG
Phenotype
MIM: 603464
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 262 Intron NP_001092003.2
NM_001160367.1 262 Intron NP_001153839.1
NM_052987.3 262 Intron NP_443713.2
NM_052988.4 262 Intron NP_443714.3
XM_006721308.2 262 Intron XP_006721371.1
XM_006721310.2 262 Intron XP_006721373.1
XM_011523405.2 262 Intron XP_011521707.1
XM_011523406.2 262 Intron XP_011521708.1
XM_011523407.2 262 Intron XP_011521709.1
XM_011523408.2 262 Intron XP_011521710.1
XM_011523410.2 262 Intron XP_011521712.1
XM_011523411.2 262 Intron XP_011521713.1
XM_011523412.1 262 Intron XP_011521714.1
XM_011523413.2 262 UTR 5 XP_011521715.1
XM_011523414.2 262 UTR 5 XP_011521716.1
XM_011523415.2 262 Intron XP_011521717.1
XM_011523416.2 262 Intron XP_011521718.1
XM_011523417.2 262 Intron XP_011521719.1
XM_017023806.1 262 Intron XP_016879295.1
XM_017023807.1 262 Intron XP_016879296.1
XM_017023808.1 262 Intron XP_016879297.1
XM_017023809.1 262 Intron XP_016879298.1
XM_017023810.1 262 UTR 5 XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

View Full Product Details