Product Details
- SNP ID
-
rs549876
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:176816453 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CCCCCTTCCTGGGCACTGATGTCAG[G/T]TGCGACAGGCAGCTTCAGACCCCAG
- Phenotype
-
MIM: 607869
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UNC5A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs60507208] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UNC5A
- Gene Name
- unc-5 netrin receptor A
View Full Product Details