Product Details
- SNP ID
-
rs298217
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:64382105 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGCGGCGTGCCGTGAGAGTCCTTT[C/G]ACAATCTCAGCTTTCATCTAGGTCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR124-2
PubMed Links
Gene Details
- Gene
- MIR124-2
- Gene Name
- microRNA 124-2
There are no transcripts associated with this gene.
- Gene
- MIR124-2HG
- Gene Name
- MIR124-2 host gene
There are no transcripts associated with this gene.
View Full Product Details