Product Details

SNP ID
rs591304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63575850 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAAGGATCACATATTAAACCCA[A/G]GTACCTCTGGAAGTGGGAAAGAATG
Phenotype
MIM: 613867
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G16 PubMed Links

Gene Details

Gene
PLA2G16
Gene Name
phospholipase A2 group XVI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128203.1 Intron NP_001121675.1
NM_007069.3 Intron NP_009000.2
XM_006718426.1 Intron XP_006718489.1
XM_011544741.1 Intron XP_011543043.1

View Full Product Details