Product Details

SNP ID
rs303007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25084463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACGGTAGGACAGCTCCCAAACC[C/T]ACAGGATATTCCTTACTTGCAAAAT
Phenotype
MIM: 603209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMAHP PubMed Links

Gene Details

Gene
CMAHP
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
There are no transcripts associated with this gene.

View Full Product Details