Product Details

SNP ID
rs150874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74189208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAGAGAAGAATGCCCTGATTGCC[A/G]AGGATATATTTTCAGCTTTGTGAAG
Phenotype
MIM: 603431 MIM: 615070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF4H PubMed Links

Gene Details

Gene
EIF4H
Gene Name
eukaryotic translation initiation factor 4H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022170.1 Intron NP_071496.1
NM_031992.1 Intron NP_114381.1
XM_011516556.1 Intron XP_011514858.1
XM_011516557.1 Intron XP_011514859.1
Gene
MIR590
Gene Name
microRNA 590
There are no transcripts associated with this gene.

View Full Product Details