Product Details

SNP ID
rs490687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:101639170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCAATAGGTCTAGTGCATAGCAT[C/G]ACTACAGTTATCATATTATAAATTT
Phenotype
MIM: 610014
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TM2D3 PubMed Links

Gene Details

Gene
TM2D3
Gene Name
TM2 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307960.1 Intron NP_001294889.1
NM_001308026.1 Intron NP_001294955.1
NM_025141.3 Intron NP_079417.2
NM_078474.2 Intron NP_510883.2

View Full Product Details