Product Details
- SNP ID
-
rs673308
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108311370 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAGCCTTATATTTGAAGAAGGG[A/T]GCCAAAGTATAACTGTTCCTAGTTT
- Phenotype
-
MIM: 607585
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76637316] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATM
- Gene Name
- ATM serine/threonine kinase
- Gene
- C11orf65
- Gene Name
- chromosome 11 open reading frame 65
View Full Product Details