Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_080927.3 | 5054 | UTR 3 | NP_563615.3 | ||
XM_011512419.2 | 5054 | UTR 3 | XP_011510721.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271142.1 | 5054 | Intron | NP_001258071.1 | ||
NM_001271145.1 | 5054 | Intron | NP_001258074.1 | ||
NM_001271146.1 | 5054 | Intron | NP_001258075.1 | ||
NM_001271147.1 | 5054 | Intron | NP_001258076.1 | ||
NM_001271148.1 | 5054 | Intron | NP_001258077.1 | ||
NM_001323352.1 | 5054 | Intron | NP_001310281.1 | ||
NM_001323353.1 | 5054 | Intron | NP_001310282.1 | ||
NM_001323358.1 | 5054 | Intron | NP_001310287.1 | ||
NM_001323359.1 | 5054 | Intron | NP_001310288.1 | ||
NM_001323360.1 | 5054 | Intron | NP_001310289.1 | ||
NM_001323362.1 | 5054 | Intron | NP_001310291.1 | ||
NM_001323363.1 | 5054 | Intron | NP_001310292.1 | ||
NM_001323364.1 | 5054 | Intron | NP_001310293.1 | ||
NM_001323365.1 | 5054 | Intron | NP_001310294.1 | ||
NM_001323366.1 | 5054 | Intron | NP_001310295.1 | ||
NM_001323367.1 | 5054 | Intron | NP_001310296.1 | ||
NM_001323368.1 | 5054 | Intron | NP_001310297.1 | ||
NM_006100.3 | 5054 | Intron | NP_006091.1 |