Product Details

SNP ID
rs335632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:77432260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGTAGACTACTTTCTAATATTTT[C/T]ATTTTTTAGCACCAAAAGGAGAAAA
Phenotype
MIM: 603390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE8B PubMed Links

Gene Details

Gene
PDE8B
Gene Name
phosphodiesterase 8B
There are no transcripts associated with this gene.

Gene
WDR41
Gene Name
WD repeat domain 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018268.3 2368 UTR 3 NP_060738.2
XM_005248551.4 2368 UTR 3 XP_005248608.1
XM_005248552.3 2368 Intron XP_005248609.1
XM_011543505.2 2368 UTR 3 XP_011541807.1
XM_011543507.2 2368 Intron XP_011541809.1
XM_017009606.1 2368 UTR 3 XP_016865095.1

View Full Product Details