Product Details

SNP ID
rs133381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42074604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACGGTGGGGCGGGCGGGAGGCC[C/G]GGTGTGCGGTGGCCTGGCCCCCCTA
Phenotype
MIM: 614240 MIM: 104170 MIM: 615588
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM109B PubMed Links

Gene Details

Gene
FAM109B
Gene Name
family with sequence similarity 109 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002034.2 Intron NP_001002034.2
XM_005261373.3 Intron XP_005261430.1
Gene
NAGA
Gene Name
alpha-N-acetylgalactosaminidase
There are no transcripts associated with this gene.

Gene
SMDT1
Gene Name
single-pass membrane protein with aspartate rich tail 1
There are no transcripts associated with this gene.

Gene
SNORD13P1
Gene Name
small nucleolar RNA, C/D box 13 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details