Product Details

SNP ID
rs10029074
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:39290882 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGCCTTTAATTTCCACAAAACAC[T/G]AGATACCTTTTCCATAATAAGAGTA
Phenotype
MIM: 102579 MIM: 608151
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
RFC1 PubMed Links

Gene Details

Gene
RFC1
Gene Name
replication factor C subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204747.1 Intron NP_001191676.1
NM_002913.4 Intron NP_002904.3
XM_011513730.1 Intron XP_011512032.1
XM_011513731.1 Intron XP_011512033.1
XM_017008528.1 Intron XP_016864017.1
Gene
WDR19
Gene Name
WD repeat domain 19
There are no transcripts associated with this gene.

View Full Product Details