Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002643.3 | 2786 | Intron | NP_002634.1 | ||
NM_173074.2 | 2786 | Intron | NP_775097.1 | ||
XM_005264369.2 | 2786 | Intron | XP_005264426.1 | ||
XM_011532908.2 | 2786 | Intron | XP_011531210.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_012249.3 | 2786 | UTR 3 | NP_036381.2 | ||
XM_005264229.2 | 2786 | UTR 3 | XP_005264286.1 | ||
XM_011532726.2 | 2786 | UTR 3 | XP_011531028.1 | ||
XM_011532728.2 | 2786 | UTR 3 | XP_011531030.1 | ||
XM_017003716.1 | 2786 | UTR 3 | XP_016859205.1 |