Product Details
- SNP ID
-
rs2083796
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:46844716 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGATGTTGAGAATGCCTCTGTGC[A/G]GACCCAAATCTATAGGATGTTATTT
- Phenotype
-
MIM: 602864
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WNT9B
PubMed Links
Gene Details
- Gene
- WNT9B
- Gene Name
- Wnt family member 9B
View Full Product Details