Product Details
- SNP ID
-
rs17841732
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:532930 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGCTGGTCTTGGCCTTTCTCAGAGT[G/C]CACTGAACAGGTGGCTTCTTGGGAC
- Phenotype
-
MIM: 116948
- Polymorphism
- G/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDC34
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17841731] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDC34
- Gene Name
- cell division cycle 34
View Full Product Details