Product Details

SNP ID
rs4639196
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:83948704 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCATATTAAACAAATCTTATTGGAA[C/G]CTTTGCATGTATGAAACTTCTATAA
Phenotype
MIM: 606018
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
EDIL3 PubMed Links

Gene Details

Gene
EDIL3
Gene Name
EGF like repeats and discoidin domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278642.1 Intron NP_001265571.1
NM_005711.4 Intron NP_005702.3

View Full Product Details