Product Details

SNP ID
rs8064840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35439408 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGAATGACATAAAACTTAAGTAG[G/T]TATGCTATAGAGTAATGCATACAGC
Phenotype
MIM: 614955 MIM: 614957
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLFN12 PubMed Links

Gene Details

Gene
SLFN12
Gene Name
schlafen family member 12
There are no transcripts associated with this gene.

Gene
SLFN13
Gene Name
schlafen family member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144682.5 4157 UTR 3 NP_653283.3
XM_005257922.2 4157 Intron XP_005257979.1
XM_011524383.2 4157 UTR 3 XP_011522685.1
XM_017024232.1 4157 UTR 3 XP_016879721.1
XM_017024233.1 4157 UTR 3 XP_016879722.1

View Full Product Details