Product Details

SNP ID
rs10888439
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:151839556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCCTCCCTTTGCCAGGCTGCC[C/T]CGGCTCAAGCTTCTCTGGCCTGAGC
Phenotype
MIM: 602943 MIM: 615653
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2CD4D PubMed Links

Gene Details

Gene
C2CD4D
Gene Name
C2 calcium dependent domain containing 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136003.1 Intron NP_001129475.1
XM_011509055.2 Intron XP_011507357.1
XM_016999989.1 Intron XP_016855478.1
Gene
LOC100132111
Gene Name
uncharacterized LOC100132111
There are no transcripts associated with this gene.

Gene
RORC
Gene Name
RAR related orphan receptor C
There are no transcripts associated with this gene.

Gene
THEM5
Gene Name
thioesterase superfamily member 5
There are no transcripts associated with this gene.

View Full Product Details