Product Details

SNP ID
rs6829716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48485797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATTGCAATATTTTATATGGCTT[A/C]TAAAGCTAGTGAAAAGATAGGGGAT
Phenotype
MIM: 607520
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC10A4 PubMed Links

Gene Details

Gene
SLC10A4
Gene Name
solute carrier family 10 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152679.3 Intron NP_689892.1
Gene
ZAR1
Gene Name
zygote arrest 1
There are no transcripts associated with this gene.

View Full Product Details