Product Details

SNP ID
rs11782652
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:81741409 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGTCATAGGATTTACACAGTTACA[G/A]GGAATTATGGCAAGAGAGTACTGTG
Phenotype
MIM: 610899
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CHMP4C PubMed Links

Gene Details

Gene
CHMP4C
Gene Name
charged multivesicular body protein 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152284.3 Intron NP_689497.1

View Full Product Details