Product Details

SNP ID
rs3738128
Assay Type
Validated
NCBI dbSNP Submissions
26
Location
Chr.1:20499992 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAGGGCCAGTGGCGAAACCAGGAGG[C/G]CTGCCTGACCAACTGCCCTTCACAT
Phenotype
MIM: 612037
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MUL1 PubMed Links

Gene Details

Gene
MUL1
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024544.2 2597 UTR 3 NP_078820.2
XM_011542137.2 2597 UTR 3 XP_011540439.1

View Full Product Details